Product Details

SNP ID
rs201971834
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:24719789 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGTGGCATAGACAATATGTTAAAC[A/G]CTGTACTGTATCTGGGCTCAAGCCA
Phenotype
MIM: 610314
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PIWIL3 PubMed Links

Gene Details

Gene
PIWIL3
Gene Name
piwi like RNA-mediated gene silencing 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001008496.3 2912 Missense Mutation CGT,TGT R831C NP_001008496.2
NM_001255975.1 2912 Missense Mutation CGT,TGT R822C NP_001242904.1

View Full Product Details