Product Details

SNP ID
rs200977899
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219246705 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGGAAGGGTTACATGATGGACACTT[C/G]TACGCCCTGAAGCGAATCCTGTGTC
Phenotype
MIM: 604719 MIM: 191110
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
GLB1L PubMed Links

Gene Details

Gene
GLB1L
Gene Name
galactosidase beta 1 like
There are no transcripts associated with this gene.

Gene
STK16
Gene Name
serine/threonine kinase 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011512067.2 526 Missense Mutation TTC,TTG F45L XP_011510369.1
XM_017005160.1 526 UTR 5 XP_016860649.1
Gene
TUBA4A
Gene Name
tubulin alpha 4a
There are no transcripts associated with this gene.

Gene
TUBA4B
Gene Name
tubulin alpha 4b
There are no transcripts associated with this gene.

View Full Product Details