Product Details

SNP ID
rs200867765
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27583127 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GAGAGGGAGTGATGTCTTCCAGACT[C/T]GGTGCTGTACCCGCCGTGAGTTTCT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
C2orf16 PubMed Links

Gene Details

Gene
C2orf16
Gene Name
chromosome 2 open reading frame 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032266.3 159 Intron NP_115642.3
Gene
ZNF512
Gene Name
zinc finger protein 512
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271286.1 159 Silent Mutation CTC,CTT L5L NP_001258215.1
NM_001271287.1 159 UTR 5 NP_001258216.1
NM_001271288.1 159 UTR 5 NP_001258217.1
NM_001271289.1 159 UTR 5 NP_001258218.1
NM_001271318.1 159 UTR 5 NP_001258247.1
NM_032434.3 159 Silent Mutation CTC,CTT L5L NP_115810.2

View Full Product Details