Product Details

SNP ID
rs199613963
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:28404252 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCATACCCTCGCTCGCACCCCTACA[G/T]CCCCCTGCCGGGCCTGGCCTCTGTC
Phenotype
MIM: 601575
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
FLJ31356 PubMed Links

Gene Details

Gene
FLJ31356
Gene Name
uncharacterized protein FLJ31356
There are no transcripts associated with this gene.

Gene
FOSL2
Gene Name
FOS like 2, AP-1 transcription factor subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005253.3 1210 Missense Mutation AGC,ATC S83I NP_005244.1
XM_005264231.3 1210 Missense Mutation AGC,ATC S83I XP_005264288.1
XM_006711976.2 1210 Missense Mutation AGC,ATC S83I XP_006712039.1
XM_006711977.3 1210 Missense Mutation AGC,ATC S44I XP_006712040.1
XM_017003737.1 1210 Missense Mutation AGC,ATC S44I XP_016859226.1

View Full Product Details