Product Details

SNP ID
rs199885480
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:219574878 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CAGCCTCCAATAGCTCTGAGCCCCT[C/G]CTAGGCCTGCTGGCACTGTCACCGG
Phenotype
MIM: 147380 MIM: 610991
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
INHA PubMed Links

Gene Details

Gene
INHA
Gene Name
inhibin alpha subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002191.3 596 Silent Mutation CTC,CTG L151L NP_002182.1
Gene
OBSL1
Gene Name
obscurin like 1
There are no transcripts associated with this gene.

View Full Product Details