Product Details
- SNP ID
-
rs200026743
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:53866816 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGAGCTGGGGCATCCAGGTGGGAAC[A/G]TCGTAAGGACTAGAAAGAACACATG
- Phenotype
-
MIM: 606704
MIM: 607705
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GPR75
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs1049894] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GPR75
- Gene Name
- G protein-coupled receptor 75
There are no transcripts associated with this gene.
- Gene
- GPR75-ASB3
- Gene Name
- GPR75-ASB3 readthrough
There are no transcripts associated with this gene.
- Gene
- PSME4
- Gene Name
- proteasome activator subunit 4
View Full Product Details