Product Details

SNP ID
rs199932153
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:70259374 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCCCTCATAGCGATTATTGGAGCC[A/G]GAATTGGTGGCACTTCAGCAGCCTA
Phenotype
MIM: 610995
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR1285-2 PubMed Links

Gene Details

Gene
MIR1285-2
Gene Name
microRNA 1285-2
There are no transcripts associated with this gene.

Gene
PCYOX1
Gene Name
prenylcysteine oxidase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_016297.3 193 Missense Mutation AGA,GGA R43G NP_057381.3

View Full Product Details