Product Details

SNP ID
rs200410805
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:166198848 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATTAACATTATCAAAAGCCATAT[C/G]TTTTTTATTGAGTAAATCATCATCT
Phenotype
MIM: 603415
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
LOC101929680 PubMed Links

Gene Details

Gene
LOC101929680
Gene Name
uncharacterized LOC101929680
There are no transcripts associated with this gene.

Gene
SCN9A
Gene Name
sodium voltage-gated channel alpha subunit 9
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002977.3 6066 Missense Mutation CAT,GAT H1920D NP_002968.1
XM_005246757.2 6066 Missense Mutation CAT,GAT H1931D XP_005246814.1
XM_011511616.2 6066 Missense Mutation CAT,GAT H1931D XP_011509918.1
XM_011511617.2 6066 Missense Mutation CAT,GAT H1931D XP_011509919.1
XM_011511618.2 6066 Missense Mutation CAT,GAT H1920D XP_011509920.1
XM_011511619.2 6066 Intron XP_011509921.1
XM_017004668.1 6066 Missense Mutation CAT,GAT H1802D XP_016860157.1
XM_017004669.1 6066 Missense Mutation CAT,GAT H1683D XP_016860158.1

View Full Product Details