Product Details

SNP ID
rs199895914
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:142885441 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCGGCTGAACTCAAATGCCACCCAA[C/T]GGATGAGAGGGTGGCTCTCCACCTA
Phenotype
MIM: 605197
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
KYNU PubMed Links

Gene Details

Gene
KYNU
Gene Name
kynureninase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001032998.1 166 Missense Mutation ACG,ATG T25M NP_001028170.1
NM_001199241.1 166 Missense Mutation ACG,ATG T25M NP_001186170.1
NM_003937.2 166 Missense Mutation ACG,ATG T25M NP_003928.1
XM_011512102.2 166 Intron XP_011510404.1
XM_017005217.1 166 Missense Mutation ACG,ATG T25M XP_016860706.1

View Full Product Details