Product Details

SNP ID
rs200050030
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27583694 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CACAACCTTTAAGCAGCAGAGGAGC[A/G]CGAGGATCGTGGGAGCTAAGAAGTA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf16 PubMed Links

Gene Details

Gene
C2orf16
Gene Name
chromosome 2 open reading frame 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032266.3 211 Intron NP_115642.3
Gene
ZNF512
Gene Name
zinc finger protein 512
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271286.1 211 Missense Mutation ACG,GCG T23A NP_001258215.1
NM_001271287.1 211 Intron NP_001258216.1
NM_001271288.1 211 Intron NP_001258217.1
NM_001271289.1 211 UTR 5 NP_001258218.1
NM_001271318.1 211 UTR 5 NP_001258247.1
NM_032434.3 211 Missense Mutation ACG,GCG T23A NP_115810.2

View Full Product Details