Product Details

SNP ID
rs201730263
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:47521331 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GATGACGTTGAAGAGCGAGTAAATG[C/G]AGCACACGCCGAGCAGGATGAAGAG
Phenotype
MIM: 607366 MIM: 609309
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
HCG2040054 PubMed Links

Gene Details

Gene
HCG2040054
Gene Name
uncharacterized LOC644093
There are no transcripts associated with this gene.

Gene
KCNK12
Gene Name
potassium two pore domain channel subfamily K member 12
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022055.1 869 Missense Mutation TCC,TGC S290C NP_071338.1
Gene
MSH2
Gene Name
mutS homolog 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000251.2 869 Intron NP_000242.1
NM_001258281.1 869 Intron NP_001245210.1
XM_005264332.3 869 Intron XP_005264389.2
XM_011532867.1 869 Intron XP_011531169.1

View Full Product Details