Product Details

SNP ID
rs201399983
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:129543242 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCATGGCTCCTGGGAGCGTCACCAG[C/T]GACATCTCACCCTCCTCGACTTCCA
Phenotype
MIM: 180380
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
H1FOO PubMed Links

Gene Details

Gene
H1FOO
Gene Name
H1 histone family member O, oocyte specific
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001308262.1 Intron NP_001295191.1
NM_153833.2 Intron NP_722575.1
XM_017005733.1 Intron XP_016861222.1
Gene
RHO
Gene Name
rhodopsin
There are no transcripts associated with this gene.

View Full Product Details