Product Details
- SNP ID
-
rs200954315
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:184365206 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGGATGATGGTGAATACAACCC[C/G]ACTGATGATAGGCCTTCCAGGTGAG
- Phenotype
-
MIM: 600570
MIM: 606023
MIM: 600044
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
CLCN2
PubMed Links
Gene Details
- Gene
- CLCN2
- Gene Name
- chloride voltage-gated channel 2
There are no transcripts associated with this gene.
- Gene
- POLR2H
- Gene Name
- RNA polymerase II subunit H
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001278698.1 |
1028 |
Silent Mutation |
CCC,CCG |
P77P |
NP_001265627.1 |
NM_001278699.1 |
1028 |
Silent Mutation |
CCC,CCG |
P41P |
NP_001265628.1 |
NM_001278700.1 |
1028 |
Silent Mutation |
CCC,CCG |
P41P |
NP_001265629.1 |
NM_001278714.1 |
1028 |
Silent Mutation |
CCC,CCG |
P77P |
NP_001265643.1 |
NM_001278715.1 |
1028 |
Silent Mutation |
CCC,CCG |
P41P |
NP_001265644.1 |
NM_006232.3 |
1028 |
Silent Mutation |
CCC,CCG |
P77P |
NP_006223.2 |
XM_006713666.2 |
1028 |
Silent Mutation |
CCC,CCG |
P77P |
XP_006713729.1 |
XM_006713667.2 |
1028 |
Silent Mutation |
CCC,CCG |
P77P |
XP_006713730.1 |
XM_006713668.3 |
1028 |
Silent Mutation |
CCC,CCG |
P77P |
XP_006713731.1 |
XM_006713670.2 |
1028 |
Silent Mutation |
CCC,CCG |
P41P |
XP_006713733.1 |
XM_017006636.1 |
1028 |
Silent Mutation |
CCC,CCG |
P77P |
XP_016862125.1 |
XM_017006637.1 |
1028 |
Silent Mutation |
CCC,CCG |
P41P |
XP_016862126.1 |
- Gene
- THPO
- Gene Name
- thrombopoietin
There are no transcripts associated with this gene.
View Full Product Details