Product Details
- SNP ID
-
rs201358681
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:10286799 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CCCTGACAGCTTGATTCCAACATCA[A/G]AGGGGGCGTTGAACTAGGAGGCAGG
- Phenotype
-
MIM: 605353
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
GHRL
PubMed Links
Gene Details
- Gene
- GHRL
- Gene Name
- ghrelin/obestatin prepropeptide
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001134941.2 |
1107 |
Missense Mutation |
TCT,TTT |
S79F |
NP_001128413.1 |
NM_001134944.1 |
1107 |
Missense Mutation |
TCT,TTT |
S68F |
NP_001128416.1 |
NM_001134945.1 |
1107 |
Missense Mutation |
TCT,TTT |
S67F |
NP_001128417.1 |
NM_001134946.1 |
1107 |
Missense Mutation |
TCT,TTT |
S29F |
NP_001128418.1 |
NM_001302821.1 |
1107 |
Missense Mutation |
TCT,TTT |
S80F |
NP_001289750.1 |
NM_001302822.1 |
1107 |
Missense Mutation |
TCT,TTT |
S80F |
NP_001289751.1 |
NM_001302823.1 |
1107 |
Missense Mutation |
TCT,TTT |
S79F |
NP_001289752.1 |
NM_001302824.1 |
1107 |
Missense Mutation |
TCT,TTT |
S80F |
NP_001289753.1 |
NM_001302825.1 |
1107 |
Missense Mutation |
TCT,TTT |
S80F |
NP_001289754.1 |
NM_016362.4 |
1107 |
Missense Mutation |
TCT,TTT |
S80F |
NP_057446.1 |
XM_017006612.1 |
1107 |
Missense Mutation |
TCT,TTT |
S80F |
XP_016862101.1 |
XM_017006613.1 |
1107 |
Missense Mutation |
TCT,TTT |
S79F |
XP_016862102.1 |
- Gene
- GHRLOS
- Gene Name
- ghrelin opposite strand/antisense RNA
There are no transcripts associated with this gene.
- Gene
- LINC00852
- Gene Name
- long intergenic non-protein coding RNA 852
There are no transcripts associated with this gene.
- Gene
- TATDN2
- Gene Name
- TatD DNase domain containing 2
There are no transcripts associated with this gene.
View Full Product Details