Product Details

SNP ID
rs200247671
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:154337937 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AAATCTGTTTTCTTTCCTGGAGTAA[C/T]GTAGGGATGGAGATCTGACTTGTGT
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
GPR149 PubMed Links

Gene Details

Gene
GPR149
Gene Name
G protein-coupled receptor 149
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001038705.1 2058 Missense Mutation NP_001033794.1

View Full Product Details