Product Details
- SNP ID
-
rs200917924
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:112540954 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- ATTATGAAAGCTGTTATCTCTTTCT[A/G]TCTGGAGTAAGAAATAGATCCTAAA
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ZGRF1
PubMed Links
Gene Details
- Gene
- ZGRF1
- Gene Name
- zinc finger GRF-type containing 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_018392.4 |
5937 |
Missense Mutation |
ACA,ATA |
T1926I |
NP_060862.3 |
XM_005263115.3 |
5937 |
Missense Mutation |
ACA,ATA |
T1926I |
XP_005263172.1 |
XM_011532091.2 |
5937 |
Missense Mutation |
ACA,ATA |
T1909I |
XP_011530393.1 |
XM_011532092.2 |
5937 |
Missense Mutation |
ACA,ATA |
T1906I |
XP_011530394.1 |
XM_011532093.2 |
5937 |
Missense Mutation |
ACA,ATA |
T1868I |
XP_011530395.1 |
XM_011532094.2 |
5937 |
Missense Mutation |
ACA,ATA |
T1840I |
XP_011530396.1 |
XM_011532096.2 |
5937 |
Intron |
|
|
XP_011530398.1 |
XM_011532097.2 |
5937 |
Missense Mutation |
ACA,ATA |
T975I |
XP_011530399.1 |
XM_011532098.2 |
5937 |
Missense Mutation |
ACA,ATA |
T883I |
XP_011530400.1 |
XM_011532099.2 |
5937 |
Missense Mutation |
ACA,ATA |
T883I |
XP_011530401.1 |
XM_017008369.1 |
5937 |
Missense Mutation |
ACA,ATA |
T1906I |
XP_016863858.1 |
XM_017008370.1 |
5937 |
Missense Mutation |
ACA,ATA |
T1851I |
XP_016863859.1 |
XM_017008371.1 |
5937 |
Missense Mutation |
ACA,ATA |
T1831I |
XP_016863860.1 |
XM_017008372.1 |
5937 |
Missense Mutation |
ACA,ATA |
T676I |
XP_016863861.1 |
XM_017008373.1 |
5937 |
Missense Mutation |
ACA,ATA |
T676I |
XP_016863862.1 |
View Full Product Details