Product Details

SNP ID
rs200999022
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:102632146 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCCAAGGGTAAAATAATATAGTTC[A/G]TGTCTTCTCAGTCATGAGGAAACCA
Phenotype
MIM: 609489
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MANBA PubMed Links

Gene Details

Gene
MANBA
Gene Name
mannosidase beta
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005908.3 1786 Nonsense Mutation CGA,TGA R851* NP_005899.3
XM_011531966.2 1786 Nonsense Mutation CGA,TGA R436* XP_011530268.1
XM_017008203.1 1786 Nonsense Mutation CGA,TGA R730* XP_016863692.1
XM_017008204.1 1786 Nonsense Mutation CGA,TGA R635* XP_016863693.1
XM_017008205.1 1786 Nonsense Mutation CGA,TGA R449* XP_016863694.1

View Full Product Details