Product Details

SNP ID
rs200853662
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:138182640 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CGGTCCCTGGCGCTGATCTTCAATA[A/G]CCTCCAAGGCCAACTTCATCCAACA
Phenotype
MIM: 602848 MIM: 603462 MIM: 605664
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
BRD8 PubMed Links

Gene Details

Gene
BRD8
Gene Name
bromodomain containing 8
There are no transcripts associated with this gene.

Gene
CDC23
Gene Name
cell division cycle 23
There are no transcripts associated with this gene.

Gene
KIF20A
Gene Name
kinesin family member 20A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005733.2 1065 Missense Mutation AAC,AGC N190S NP_005724.1

View Full Product Details