Product Details
- SNP ID
-
rs201385839
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:156644527 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- CACTCATCTAAGAAAGATAACTTGG[A/C]AGTCAATGCAGTTGCTTTACAAGAT
- Phenotype
-
MIM: 610242
MIM: 610241
- Polymorphism
- A/C, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
C7orf13
PubMed Links
Gene Details
- Gene
- C7orf13
- Gene Name
- chromosome 7 open reading frame 13
There are no transcripts associated with this gene.
- Gene
- RNF32
- Gene Name
- ring finger protein 32
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001184996.1 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
NP_001171925.1 |
NM_001184997.1 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
NP_001171926.1 |
NM_001308273.1 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
NP_001295202.1 |
NM_001308274.1 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
NP_001295203.1 |
NM_030936.3 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
NP_112198.1 |
XM_005249522.4 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_005249579.1 |
XM_011515804.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514106.1 |
XM_011515805.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514107.1 |
XM_011515806.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514108.1 |
XM_011515807.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514109.1 |
XM_011515808.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514110.1 |
XM_011515809.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514111.1 |
XM_011515810.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514112.1 |
XM_011515811.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514113.1 |
XM_011515812.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514114.1 |
XM_011515813.2 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_011514115.1 |
XM_017011753.1 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_016867242.1 |
XM_017011754.1 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_016867243.1 |
XM_017011755.1 |
284 |
Missense Mutation |
GAA,GCA |
E15A |
XP_016867244.1 |
View Full Product Details