Product Details
- SNP ID
-
rs201218381
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:87194728 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- TAGAAGAACAAGTTGATCAAACAAT[C/T]GATGATGAAACAATACTTATCGTTC
- Phenotype
-
MIM: 608491
MIM: 616993
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
DMTF1
PubMed Links
Gene Details
- Gene
- DMTF1
- Gene Name
- cyclin D binding myb like transcription factor 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_001142326.1 |
3587 |
Silent Mutation |
ATC,ATT |
I603I |
NP_001135798.1 |
NM_001142327.1 |
3587 |
Silent Mutation |
ATC,ATT |
I691I |
NP_001135799.1 |
NM_021145.3 |
3587 |
Silent Mutation |
ATC,ATT |
I691I |
NP_066968.3 |
XM_011516735.2 |
3587 |
Silent Mutation |
ATC,ATT |
I691I |
XP_011515037.1 |
XM_011516737.1 |
3587 |
Silent Mutation |
ATC,ATT |
I691I |
XP_011515039.1 |
XM_011516739.2 |
3587 |
Silent Mutation |
ATC,ATT |
I650I |
XP_011515041.1 |
XM_017012865.1 |
3587 |
Silent Mutation |
ATC,ATT |
I691I |
XP_016868354.1 |
XM_017012866.1 |
3587 |
Silent Mutation |
ATC,ATT |
I650I |
XP_016868355.1 |
XM_017012867.1 |
3587 |
Silent Mutation |
ATC,ATT |
I650I |
XP_016868356.1 |
XM_017012868.1 |
3587 |
Intron |
|
|
XP_016868357.1 |
XM_017012869.1 |
3587 |
Silent Mutation |
ATC,ATT |
I632I |
XP_016868358.1 |
XM_017012870.1 |
3587 |
Intron |
|
|
XP_016868359.1 |
XM_017012871.1 |
3587 |
Silent Mutation |
ATC,ATT |
I425I |
XP_016868360.1 |
XM_017012872.1 |
3587 |
Silent Mutation |
ATC,ATT |
I425I |
XP_016868361.1 |
XM_017012873.1 |
3587 |
Silent Mutation |
ATC,ATT |
I425I |
XP_016868362.1 |
XM_017012874.1 |
3587 |
Silent Mutation |
ATC,ATT |
I425I |
XP_016868363.1 |
XM_017012875.1 |
3587 |
Silent Mutation |
ATC,ATT |
I425I |
XP_016868364.1 |
XM_017012876.1 |
3587 |
Silent Mutation |
ATC,ATT |
I425I |
XP_016868365.1 |
XM_017012877.1 |
3587 |
Intron |
|
|
XP_016868366.1 |
XM_017012878.1 |
3587 |
Intron |
|
|
XP_016868367.1 |
- Gene
- TMEM243
- Gene Name
- transmembrane protein 243
View Full Product Details