Product Details

SNP ID
rs199815125
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:73976409 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGTCTCCCGGGCGTCCTCCAGCA[A/G]CGGGCCTTCGGGGCCGGTAGCCGGC
Phenotype
MIM: 600788 MIM: 612418
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TCEB1 PubMed Links

Gene Details

Gene
TCEB1
Gene Name
transcription elongation factor B subunit 1
There are no transcripts associated with this gene.

Gene
TMEM70
Gene Name
transmembrane protein 70
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040613.2 268 Missense Mutation AAC,AGC N43S NP_001035703.1
NM_017866.5 268 Missense Mutation AAC,AGC N43S NP_060336.3

View Full Product Details