Product Details

SNP ID
rs199738743
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:42399695 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGACTCTTGATACCATATTTGTACC[G/T]AACACAGGGTAATTATTCAAATCCC
Phenotype
MIM: 605417 MIM: 610029
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
DKK4 PubMed Links

Gene Details

Gene
DKK4
Gene Name
dickkopf WNT signaling pathway inhibitor 4
There are no transcripts associated with this gene.

Gene
VDAC3
Gene Name
voltage dependent anion channel 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135694.2 528 Silent Mutation CCG,CCT P106P NP_001129166.1
NM_005662.6 528 Silent Mutation CCG,CCT P105P NP_005653.3
XM_006716394.1 528 Silent Mutation CCG,CCT P73P XP_006716457.1

View Full Product Details