Product Details

SNP ID
rs200902701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:33240233 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CATGGCCGTCCGCCAGTGGGTAATC[A/G]CCCTGGCCTTGGCTGCCCTCCTTGT
Phenotype
MIM: 613929
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SPINK4 PubMed Links

Gene Details

Gene
SPINK4
Gene Name
serine peptidase inhibitor, Kazal type 4
There are no transcripts associated with this gene.

View Full Product Details