Product Details

SNP ID
rs199615228
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133066564 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTTCGGGATCAGCACATGGCCAT[C/T]GCTTGGGTGAAGAGGAATATCGCGG
Phenotype
MIM: 114840 MIM: 604890
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CEL PubMed Links

Gene Details

Gene
CEL
Gene Name
carboxyl ester lipase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001807.4 598 Silent Mutation ATC,ATT I194I NP_001798.2
Gene
GTF3C5
Gene Name
general transcription factor IIIC subunit 5
There are no transcripts associated with this gene.

Gene
LOC100996574
Gene Name
uncharacterized LOC100996574
There are no transcripts associated with this gene.

View Full Product Details