Product Details

SNP ID
rs201969467
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:128051254 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGTACCTTTGGAAGCCAGCTGTTCC[A/G]CTTCCTTCATGAGCCTTTCCTCCAG
Phenotype
MIM: 300487
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ACTRT1 PubMed Links

Gene Details

Gene
ACTRT1
Gene Name
actin related protein T1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_138289.3 1150 Missense Mutation GCG,GTG A318V NP_612146.1

View Full Product Details