Product Details

SNP ID
hCV192282916
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154506862 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TACAGCCTTCCATCTCCAGCGCCTC[A/G]GGCCAGCCTTCGTACTTGTTGCTGT
Phenotype
MIM: 300492
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
FAM3A PubMed Links

Gene Details

Gene
FAM3A
Gene Name
family with sequence similarity 3 member A
There are no transcripts associated with this gene.

View Full Product Details