Product Details

SNP ID
rs200891603
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:70426930 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCTGGTCATCTACTTGGCCCCCCT[A/C]TGCATCTCCTCACCCTGCATCATGG
Phenotype
MIM: 300521
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
GDPD2 PubMed Links

Gene Details

Gene
GDPD2
Gene Name
glycerophosphodiester phosphodiesterase domain containing 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171191.1 498 Silent Mutation CTA,CTC L128L NP_001164662.1
NM_001171192.1 498 Silent Mutation CTA,CTC L207L NP_001164663.1
NM_001171193.1 498 Silent Mutation CTA,CTC L128L NP_001164664.1
NM_017711.3 498 Silent Mutation CTA,CTC L207L NP_060181.2
XM_011530977.1 498 Silent Mutation CTA,CTC L80L XP_011529279.1
XM_017029614.1 498 UTR 5 XP_016885103.1
Gene
KIF4A
Gene Name
kinesin family member 4A
There are no transcripts associated with this gene.

Gene
LOC105373244
Gene Name
uncharacterized LOC105373244
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
XM_011531081.2 498 Intron XP_011529383.1
XM_011531082.2 498 Intron XP_011529384.1

View Full Product Details