Product Details

SNP ID
rs200815004
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:65518021 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGCCATGCAGAGCCCTGCAAAGC[G/T]CCTCTTTTCTGGGCCAGAAGCCTAG
Phenotype
MIM: 300964
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
LAS1L PubMed Links

Gene Details

Gene
LAS1L
Gene Name
LAS1 like, ribosome biogenesis factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001170649.1 1918 Silent Mutation GGA,GGC G614G NP_001164120.1
NM_001170650.1 1918 Silent Mutation GGA,GGC G572G NP_001164121.1
NM_031206.4 1918 Silent Mutation GGA,GGC G631G NP_112483.1
XM_005262301.1 1918 Silent Mutation GGA,GGC G631G XP_005262358.1
XM_005262304.1 1918 Intron XP_005262361.1
XM_005262305.3 1918 Intron XP_005262362.1
XM_005262306.3 1918 Intron XP_005262363.1
XM_011531045.1 1918 Silent Mutation GGA,GGC G589G XP_011529347.1
XM_011531046.1 1918 Intron XP_011529348.1
XM_017029877.1 1918 Silent Mutation GGA,GGC G614G XP_016885366.1
XM_017029878.1 1918 Intron XP_016885367.1
XM_017029879.1 1918 Silent Mutation GGA,GGC G329G XP_016885368.1
XM_017029880.1 1918 Silent Mutation GGA,GGC G312G XP_016885369.1

View Full Product Details