Product Details
- SNP ID
-
rs200478641
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:77508395 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAAGGTCCTGGATTTTTGCTTCTCA[C/T]TGGGGGTGGTGCACGCTGTAATGGT
- Phenotype
-
MIM: 300504
MIM: 300032
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
ATRX
PubMed Links
Gene Details
- Gene
- ATRX
- Gene Name
- ATRX, chromatin remodeler
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_000489.4 |
7717 |
Missense Mutation |
ATG,GTG |
M2479V |
NP_000480.3 |
NM_138270.3 |
7717 |
Missense Mutation |
ATG,GTG |
M2441V |
NP_612114.2 |
XM_005262153.4 |
7717 |
Missense Mutation |
ATG,GTG |
M2478V |
XP_005262210.2 |
XM_005262154.4 |
7717 |
Missense Mutation |
ATG,GTG |
M2450V |
XP_005262211.2 |
XM_005262155.4 |
7717 |
Missense Mutation |
ATG,GTG |
M2440V |
XP_005262212.2 |
XM_005262156.4 |
7717 |
Missense Mutation |
ATG,GTG |
M2424V |
XP_005262213.2 |
XM_005262157.4 |
7717 |
Missense Mutation |
ATG,GTG |
M2411V |
XP_005262214.2 |
XM_006724666.3 |
7717 |
Missense Mutation |
ATG,GTG |
M2440V |
XP_006724729.1 |
XM_006724667.3 |
7717 |
Missense Mutation |
ATG,GTG |
M2386V |
XP_006724730.1 |
XM_006724668.2 |
7717 |
Intron |
|
|
XP_006724731.1 |
XM_017029601.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2449V |
XP_016885090.1 |
XM_017029602.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2439V |
XP_016885091.1 |
XM_017029603.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2423V |
XP_016885092.1 |
XM_017029604.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2412V |
XP_016885093.1 |
XM_017029605.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2411V |
XP_016885094.1 |
XM_017029606.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2402V |
XP_016885095.1 |
XM_017029607.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2401V |
XP_016885096.1 |
XM_017029608.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2385V |
XP_016885097.1 |
XM_017029609.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2373V |
XP_016885098.1 |
XM_017029610.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2372V |
XP_016885099.1 |
XM_017029611.1 |
7717 |
Missense Mutation |
ATG,GTG |
M2357V |
XP_016885100.1 |
View Full Product Details