Product Details
- SNP ID
-
rs1047584
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:187370868 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TGCCACACGTACTGGGAGCACTGGA[C/T]ATCCCAGGGTCAGGTGCGTATGAGG
- Phenotype
-
MIM: 609350
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
RTP4
PubMed Links
Gene Details
- Gene
- RTP4
- Gene Name
- receptor transporter protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_022147.2 |
298 |
Missense Mutation |
ACA,ATA |
T79I |
NP_071430.2 |
View Full Product Details