Product Details

SNP ID
rs12478841
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:27588855 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAGTTTTCTGATTTATGAACATGCT[A/G]TATTTTCCCCTTTAAGTCTTCTTAA
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
C2orf16 PubMed Links

Gene Details

Gene
C2orf16
Gene Name
chromosome 2 open reading frame 16
There are no transcripts associated with this gene.

Gene
ZNF512
Gene Name
zinc finger protein 512
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001271286.1 Intron NP_001258215.1
NM_001271287.1 Intron NP_001258216.1
NM_001271288.1 Intron NP_001258217.1
NM_001271289.1 Intron NP_001258218.1
NM_001271318.1 Intron NP_001258247.1
NM_032434.3 Intron NP_115810.2

View Full Product Details