Product Details

SNP ID
rs12614550
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:15938103 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTCTCCATGTACAAAGTCTTTTATC[C/T]TTCTACTTACAAATGAAGGTAACCA
Phenotype
MIM: 164840 MIM: 605374 MIM: 615968
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
MYCN PubMed Links

Gene Details

Gene
MYCN
Gene Name
v-myc avian myelocytomatosis viral oncogene neuroblastoma derived homolog
There are no transcripts associated with this gene.

Gene
MYCNOS
Gene Name
MYCN opposite strand
There are no transcripts associated with this gene.

Gene
MYCNUT
Gene Name
MYCN upstream transcript (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details