Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_017925.6 | Intron | NP_060395.5 | |||
XM_006716804.3 | Intron | XP_006716867.1 | |||
XM_006716805.3 | Intron | XP_006716868.1 | |||
XM_006716807.3 | Intron | XP_006716870.1 | |||
XM_011517959.2 | Intron | XP_011516261.1 | |||
XM_017014917.1 | Intron | XP_016870406.1 | |||
XM_017014918.1 | Intron | XP_016870407.1 | |||
XM_017014919.1 | Intron | XP_016870408.1 | |||
XM_017014920.1 | Intron | XP_016870409.1 |