Product Details
- SNP ID
-
rs6675902
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
33
- Location
-
Chr.1:47105832 on Build GRCh38
- Set Membership
-
Validated
- Context Sequence [VIC/FAM]
- GCACTCTGGATATAAATACAGTCCA[C/T]GTCTCTACCAAATTTTCACAGATGT
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
CYP4Z1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs10567873] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- CYP4Z1
- Gene Name
- cytochrome P450 family 4 subfamily Z member 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_178134.2 |
|
Intron |
|
|
NP_835235.1 |
View Full Product Details