Product Details

SNP ID
rs1042262
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.12:20369473 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCCGGAGCTCTCGGAAACTTTCCTC[C/T]GCGCTGTGCGCGGGCTCCCTGTCCT
Phenotype
MIM: 123805
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
PDE3A PubMed Links

Gene Details

Gene
PDE3A
Gene Name
phosphodiesterase 3A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000921.4 229 Silent Mutation TCC,TCT S63S NP_000912.3
NM_001244683.1 229 Intron NP_001231612.1
XM_017019420.1 229 Silent Mutation TCC,TCT S63S XP_016874909.1
XM_017019421.1 229 Intron XP_016874910.1

View Full Product Details