Product Details

SNP ID
rs2011560
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.8:8319138 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CGCAGTCCTGCTCCTGGGCAGGGGG[C/G]TGTGTGGGCAGGGCAGGCACAGGGT
Phenotype
Polymorphism
C/G, Transversion Substitution
Allele Nomenclature
Literature Links
SGK223 PubMed Links

Gene Details

Gene
SGK223
Gene Name
homolog of rat pragma of Rnd2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001080826.2 3912 Silent Mutation CAC,CAG H1079Q NP_001074295.2
XM_005272369.4 3912 Missense Mutation CAC,CAG H1079Q XP_005272426.2
XM_005272370.4 3912 Missense Mutation CAC,CAG H1079Q XP_005272427.2
XM_011543813.2 3912 Intron XP_011542115.1

View Full Product Details