Product Details
- SNP ID
-
rs1791627
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:52361313 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- AACACTTGCGAAGAGCACTCAGCTC[A/T]GCTTTCTCTAACACCTGTCATGGCC
- Phenotype
-
MIM: 602767
- Polymorphism
- A/T, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
KRT85
PubMed Links
Gene Details
- Gene
- KRT85
- Gene Name
- keratin 85
View Full Product Details