Product Details

SNP ID
rs1324218
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:100912208 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TTACAAAGGAGGGTTGGAGATTGCA[A/G]GCAGCTTTTACTTATCTATGCTGCC
Phenotype
MIM: 610348
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLF2 PubMed Links

Gene Details

Gene
SLF2
Gene Name
SMC5-SMC6 complex localization factor 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001136123.1 1979 Intron NP_001129595.1
NM_001243770.1 1979 Intron NP_001230699.1
NM_018121.3 1979 Intron NP_060591.3
XM_005269965.2 1979 Intron XP_005270022.1
XM_011539944.2 1979 UTR 5 XP_011538246.1

View Full Product Details