Product Details

SNP ID
rs2300548
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:64995209 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGATGAAGAGCAGACACTAAAGCCA[A/G]CCACCTGGGTTTGAAGCCCAGCTCT
Phenotype
MIM: 600229
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
SLC1A4 PubMed Links

Gene Details

Gene
SLC1A4
Gene Name
solute carrier family 1 member 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001193493.1 Intron NP_001180422.1
NM_003038.4 Intron NP_003029.2
XM_006712079.2 Intron XP_006712142.1

View Full Product Details