Product Details
- SNP ID
-
rs2111699
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.14:77322254 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- ATCAGAATGCATATTTGAATTGGAA[A/G]CACAGGTCTTCTAGGAGTGCTGATG
- Phenotype
-
MIM: 603758
MIM: 607439
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GSTZ1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs148555154] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GSTZ1
- Gene Name
- glutathione S-transferase zeta 1
- Gene
- POMT2
- Gene Name
- protein O-mannosyltransferase 2
There are no transcripts associated with this gene.
View Full Product Details