Product Details

SNP ID
rs2231182
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.18:905134 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTCTTTCGCTTGGCATCGCGTCAG[A/G]GGAGTTAGCTTTCCTTCAGCCGGGT
Phenotype
MIM: 102980
Polymorphism
A/G, Transition Substitution
Allele Nomenclature
Literature Links
ADCYAP1 PubMed Links

Gene Details

Gene
ADCYAP1
Gene Name
adenylate cyclase activating polypeptide 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001099733.1 Intron NP_001093203.1
NM_001117.4 Intron NP_001108.2
XM_005258081.3 Intron XP_005258138.1

View Full Product Details