Product Details
- SNP ID
-
rs2232117
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:33727495 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GGTTTTCCCCACAAGCCACAAGGCC[C/T]GAGGCACGCCTCCTGCCCTGCCTCC
- Phenotype
-
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
FLJ20712
PubMed Links
Gene Details
- Gene
- FLJ20712
- Gene Name
- uncharacterized FLJ20712
View Full Product Details