Product Details

SNP ID
rs2070824
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:154506931 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCTGTGGGGAGGGGAGCAGAAAGTC[A/C]TGACTTTGGCCCTCAGGGACAGGGG
Phenotype
MIM: 300492
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
FAM3A PubMed Links

Gene Details

Gene
FAM3A
Gene Name
family with sequence similarity 3 member A
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001171132.2 Intron NP_001164603.1
NM_001171133.2 Intron NP_001164604.1
NM_001171134.2 Intron NP_001164605.1
NM_001282311.1 Intron NP_001269240.1
NM_001282312.1 Intron NP_001269241.1
NM_021806.3 Intron NP_068578.2
XM_005274714.3 Intron XP_005274771.1
XM_005274716.3 Intron XP_005274773.1
XM_005277879.4 Intron XP_005277936.1
XM_006724831.2 Intron XP_006724894.1
XM_006724832.3 Intron XP_006724895.1
XM_006724833.2 Intron XP_006724896.1
XM_006724834.2 Intron XP_006724897.1
XM_011531185.2 Intron XP_011529487.1
XM_017029699.1 Intron XP_016885188.1
XM_017029700.1 Intron XP_016885189.1
XM_017029701.1 Intron XP_016885190.1
XM_017029702.1 Intron XP_016885191.1

View Full Product Details