Product Details

SNP ID
rs2282502
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.11:67066781 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GCCTGCGGCCCCTGTTCTACCCTGA[T/C]GCCAGCGTCCTGCTGCTTTGCTTCG
Phenotype
MIM: 605781
Polymorphism
T/C, Transition substitution
Allele Nomenclature
Literature Links
RHOD PubMed Links

Gene Details

Gene
RHOD
Gene Name
ras homolog family member D
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300886.1 349 Intron NP_001287815.1
NM_014578.3 349 Silent Mutation GAC,GAT D88D NP_055393.1

View Full Product Details