Product Details

SNP ID
rs2232687
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:101200170 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAAATGTGTTACTTTTTAGAATCCT[C/G]AACCTTCTCAGTATAGACAATTATC
Phenotype
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
ZNF706 PubMed Links

Gene Details

Gene
ZNF706
Gene Name
zinc finger protein 706
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001042510.1 Intron NP_001035975.1
NM_001267708.1 Intron NP_001254637.1
NM_001267709.1 Intron NP_001254638.1
NM_016096.3 Intron NP_057180.1

View Full Product Details