Product Details
- SNP ID
-
rs2233998
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141778508 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GTAAAGATGCTTCGGTTATTCTATT[C/T]CTCTGCTATTATTGCCTCAGTTATT
- Phenotype
-
MIM: 604869
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TAS2R4
PubMed Links
Gene Details
- Gene
- TAS2R4
- Gene Name
- taste 2 receptor member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016944.1 |
20 |
Missense Mutation |
TCC,TTC |
S7F |
NP_058640.1 |
View Full Product Details