Product Details
- SNP ID
-
rs2234002
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.7:141779000 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AGAAATAACACATCATTTAATATCA[A/G]TGAGGGCATCTTGTCTTTAGTGGTT
- Phenotype
-
MIM: 604869
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TAS2R4
PubMed Links
Gene Details
- Gene
- TAS2R4
- Gene Name
- taste 2 receptor member 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_016944.1 |
512 |
Missense Mutation |
AAT,AGT |
N171S |
NP_058640.1 |
View Full Product Details