Product Details
- SNP ID
-
rs2284463
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:29749309 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GTTTTACTCAGAATTTATGCAGCTG[A/G]TCACATAGGGACAGGACTGGATGAT
- Phenotype
-
MIM: 138245
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
GRIK1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs147019555] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GRIK1
- Gene Name
- glutamate ionotropic receptor kainate type subunit 1
- Gene
- GRIK1-AS1
- Gene Name
- GRIK1 antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details