Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001039140.2 | 1399 | UTR 3 | NP_001034229.1 | ||
NM_001258429.1 | 1399 | UTR 3 | NP_001245358.1 | ||
NM_001258430.1 | 1399 | UTR 3 | NP_001245359.1 | ||
XM_011529266.2 | 1399 | UTR 3 | XP_011527568.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001197327.1 | 1399 | Intron | NP_001184256.1 | ||
NM_001318322.1 | 1399 | Intron | NP_001305251.1 | ||
NM_052970.4 | 1399 | Intron | NP_443202.3 | ||
XM_011529151.2 | 1399 | Intron | XP_011527453.1 | ||
XM_017027632.1 | 1399 | Intron | XP_016883121.1 | ||
XM_017027633.1 | 1399 | Intron | XP_016883122.1 | ||
XM_017027634.1 | 1399 | Intron | XP_016883123.1 | ||
XM_017027635.1 | 1399 | Intron | XP_016883124.1 |