Product Details

SNP ID
rs2688885
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.10:50310552 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATTTAAAAAACCACAAAACAATAA[C/T]ACACAAGTCCAAAAAAATGGATGGA
Phenotype
MIM: 611573
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SGMS1 PubMed Links

Gene Details

Gene
SGMS1
Gene Name
sphingomyelin synthase 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_147156.3 Intron NP_671512.1
XM_005269675.1 Intron XP_005269732.1
XM_011539582.2 Intron XP_011537884.1
XM_011539583.2 Intron XP_011537885.1
XM_011539584.2 Intron XP_011537886.1

View Full Product Details